ADVH49B83203 AChemBlock

(R)-3-(1-aminopropyl)benzonitrile hydrochloride

  • Cas Number : 1253792-93-6
  • Molecular Weight : 196.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-3-1-aminopropyl-benzonitrile-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87480
Availability details for
Quantity
  • Availability
  • Add to Cart