ADVH49B83204 AChemBlock

(R)-1-(2,4-difluorophenyl)propan-1-amine hydrochloride

  • Cas Number : 1253792-95-8
  • Molecular Weight : 207.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-1-2-4-difluorophenyl-propan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87481
Availability details for
Quantity
  • Availability
  • Add to Cart