ADVH49B83205 AChemBlock

(R)-2-amino-3-(2-methoxyphenyl)propan-1-ol

  • Cas Number : 1255037-24-1
  • Molecular Weight : 181.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-2-amino-3-2-methoxyphenyl-propan-1-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87482
Availability details for
Quantity
  • Availability
  • Add to Cart