ADVH49B83207 AChemBlock

(S)-2-amino-3,3,3-trifluoropropan-1-ol hydrochloride

  • Cas Number : 1255946-09-8
  • Molecular Weight : 165.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-2-amino-3-3-3-trifluoropropan-1-ol-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87484
Availability details for
Quantity
  • Availability
  • Add to Cart