ADVH49B83208 AChemBlock

N-(4-chloro-3-nitrophenyl)-4-(trifluoromethyl)benzothioamide

  • Cas Number : 1257095-61-6
  • Molecular Weight : 360.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-4-chloro-3-nitrophenyl-4-trifluoromethyl-benzothioamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87485
Availability details for
Quantity
  • Availability
  • Add to Cart