ADVH49B83210 AChemBlock

5-bromo-7-fluorochroman-4-one

  • Cas Number : 1260008-29-4
  • Molecular Weight : 245.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-7-fluorochroman-4-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87493
Availability details for
Quantity
  • Availability
  • Add to Cart