ADVH49B83217 AChemBlock

7-bromo-6-fluoro-3,4-dihydronaphthalen-1(2H)-one

  • Cas Number : 1260014-75-2
  • Molecular Weight : 243.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/7-bromo-6-fluoro-3-4-dihydronaphthalen-1-2h-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87500
Availability details for
Quantity
  • Availability
  • Add to Cart