ADVH49B8321D AChemBlock

(S)-2-amino-2-(3-chloro-4-fluorophenyl)ethan-1-ol hydrochloride

  • Cas Number : 1269652-47-2
  • Molecular Weight : 226.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-2-amino-2-3-chloro-4-fluorophenyl-ethan-1-ol-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87506
Availability details for
Quantity
  • Availability
  • Add to Cart