ADVH49B83220 AChemBlock

1-(6-(trifluoromethyl)pyridin-3-yl)propan-1-amine

  • Cas Number : 1270424-42-4
  • Molecular Weight : 204.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-6-trifluoromethyl-pyridin-3-yl-propan-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87509
Availability details for
Quantity
  • Availability
  • Add to Cart