ADVH49B83225 AChemBlock

3-bromo-5-oxo-5,6,7,8-tetrahydronaphthalene-1-carboxylic acid

  • Cas Number : 1273602-53-1
  • Molecular Weight : 269.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-5-oxo-5-6-7-8-tetrahydronaphthalene-1-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87514
Availability details for
Quantity
  • Availability
  • Add to Cart