ADVH49B83229 AChemBlock

methyl 6-fluoro-3-oxo-2,3-dihydro-1H-indene-5-carboxylate

  • Cas Number : 1273651-10-7
  • Molecular Weight : 208.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-6-fluoro-3-oxo-2-3-dihydro-1h-indene-5-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87518
Availability details for
Quantity
  • Availability
  • Add to Cart