ADVH49B8322A AChemBlock

5-chloro-8-fluorochroman-4-one

  • Cas Number : 1273653-86-3
  • Molecular Weight : 200.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-chloro-8-fluorochroman-4-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87519
Availability details for
Quantity
  • Availability
  • Add to Cart