ADVH49B8322B AChemBlock

5-bromo-6-methyl-2,3-dihydro-1H-inden-1-one

  • Cas Number : 1273655-83-6
  • Molecular Weight : 225.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-6-methyl-2-3-dihydro-1h-inden-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87520
Availability details for
Quantity
  • Availability
  • Add to Cart