ADVH49B83246 AChemBlock

7-bromo-5-methoxy-3,4-dihydronaphthalen-1(2H)-one

  • Cas Number : 1336953-53-7
  • Molecular Weight : 255.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/7-bromo-5-methoxy-3-4-dihydronaphthalen-1-2h-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87547
Availability details for
Quantity
  • Availability
  • Add to Cart