ADVH49B83247 AChemBlock

2-(2-bromo-6-chlorophenyl)pyrrolidine

  • Cas Number : 1337065-54-9
  • Molecular Weight : 260.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-2-bromo-6-chlorophenyl-pyrrolidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87548
Availability details for
Quantity
  • Availability
  • Add to Cart