ADVH49B83249 AChemBlock

1-(3-chloro-5-(trifluoromethyl)phenyl)ethan-1-amine

  • Cas Number : 1337269-44-9
  • Molecular Weight : 223.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-3-chloro-5-trifluoromethyl-phenyl-ethan-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87550
Availability details for
Quantity
  • Availability
  • Add to Cart