ADVH49B8324A AChemBlock

6-bromo-1,2,3,4-tetrahydronaphthalen-1-amine

  • Cas Number : 1337523-99-5
  • Molecular Weight : 226.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-1-2-3-4-tetrahydronaphthalen-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87551
Availability details for
Quantity
  • Availability
  • Add to Cart