ADVH49B8324F AChemBlock

(R)-1-(2,3-difluorophenyl)ethan-1-amine

  • Cas Number : 1339886-04-2
  • Molecular Weight : 157.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-1-2-3-difluorophenyl-ethan-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87556
Availability details for
Quantity
  • Availability
  • Add to Cart