ADVH49B83251 AChemBlock

methyl (S)-3-amino-3-(4-hydroxyphenyl)propanoate hydrochloride

  • Cas Number : 134430-96-9
  • Molecular Weight : 231.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-s-3-amino-3-4-hydroxyphenyl-propanoate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87558
Availability details for
Quantity
  • Availability
  • Add to Cart