ADVH49B83253 AChemBlock

methyl 3-bromo-5-oxo-5,6,7,8-tetrahydronaphthalene-1-carboxylate

  • Cas Number : 1344902-57-3
  • Molecular Weight : 283.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-bromo-5-oxo-5-6-7-8-tetrahydronaphthalene-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87560
Availability details for
Quantity
  • Availability
  • Add to Cart