ADVH49B83254 AChemBlock

(S)-2-allylpiperidine hydrochloride

  • Cas Number : 1352232-87-1
  • Molecular Weight : 161.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-2-allylpiperidine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87561
Availability details for
Quantity
  • Availability
  • Add to Cart