ADVH49B83255 AChemBlock

tert-butyl 2-(6-chloropyridin-3-yl)pyrrolidine-1-carboxylate

  • Cas Number : 1352534-35-0
  • Molecular Weight : 282.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-6-chloropyridin-3-yl-pyrrolidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87562
Availability details for
Quantity
  • Availability
  • Add to Cart