ADVH49B8325A AChemBlock

methyl 3-amino-3-(2,3-dimethoxyphenyl)propanoate hydrochloride

  • Cas Number : 1369494-59-6
  • Molecular Weight : 275.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-amino-3-2-3-dimethoxyphenyl-propanoate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87567
Availability details for
Quantity
  • Availability
  • Add to Cart