ADVH49B8325C AChemBlock

(3-methylisoxazol-4-yl)methanamine hydrochloride

  • Cas Number : 1373029-26-5
  • Molecular Weight : 148.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-methylisoxazol-4-yl-methanamine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87569
Availability details for
Quantity
  • Availability
  • Add to Cart