ADVH49B83260 AChemBlock

(R)-2-(2-methoxyphenyl)pyrrolidine hydrochloride

  • Cas Number : 1381928-34-2
  • Molecular Weight : 213.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-2-2-methoxyphenyl-pyrrolidine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87573
Availability details for
Quantity
  • Availability
  • Add to Cart