ADVH49B83263 AChemBlock

(R)-2-(o-tolyl)pyrrolidine hydrochloride

  • Cas Number : 1381929-14-1
  • Molecular Weight : 197.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-2-o-tolyl-pyrrolidine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87576
Availability details for
Quantity
  • Availability
  • Add to Cart