ADVH49B83265 AChemBlock

(R)-1-(m-tolyl)ethan-1-amine

  • Cas Number : 138457-19-9
  • Molecular Weight : 135.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-1-m-tolyl-ethan-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87578
Availability details for
Quantity
  • Availability
  • Add to Cart