ADVH49B83266 AChemBlock

4-oxochromane-6-carbonitrile

  • Cas Number : 138801-92-0
  • Molecular Weight : 173.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-oxochromane-6-carbonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87579
Availability details for
Quantity
  • Availability
  • Add to Cart