ADVH49B8326A AChemBlock

(R)-1-(4-fluorophenyl)but-3-en-1-amine hydrochloride

  • Cas Number : 1391361-52-6
  • Molecular Weight : 201.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-1-4-fluorophenyl-but-3-en-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87583
Availability details for
Quantity
  • Availability
  • Add to Cart