ADVH49B8326B AChemBlock

methyl (R)-2-(1-aminoethyl)benzoate hydrochloride

  • Cas Number : 1391373-95-7
  • Molecular Weight : 215.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-r-2-1-aminoethyl-benzoate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87584
Availability details for
Quantity
  • Availability
  • Add to Cart