ADVH49B83272 AChemBlock

(S)-4-methoxy-3-(pyrrolidin-2-yl)benzoic acid hydrochloride

  • Cas Number : 1391419-55-8
  • Molecular Weight : 257.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-4-methoxy-3-pyrrolidin-2-yl-benzoic-acid-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87591
Availability details for
Quantity
  • Availability
  • Add to Cart