ADVH49B83273 AChemBlock

(S)-1-(3-chlorophenyl)-2,2,2-trifluoroethan-1-amine hydrochloride

  • Cas Number : 1391426-98-4
  • Molecular Weight : 246.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-1-3-chlorophenyl-2-2-2-trifluoroethan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87592
Availability details for
Quantity
  • Availability
  • Add to Cart