ADVH49B83276 AChemBlock

(S)-1-(2-chloro-6-fluorophenyl)propan-1-amine hydrochloride

  • Cas Number : 1391435-53-2
  • Molecular Weight : 224.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-1-2-chloro-6-fluorophenyl-propan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87595
Availability details for
Quantity
  • Availability
  • Add to Cart