ADVH49B83278 AChemBlock

(S)-2-methyl-1-(p-tolyl)propan-1-amine hydrochloride

  • Cas Number : 1391437-15-2
  • Molecular Weight : 199.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-2-methyl-1-p-tolyl-propan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87597
Availability details for
Quantity
  • Availability
  • Add to Cart