ADVH49B8327B AChemBlock

(S)-1-(4-chloro-2-methylphenyl)ethan-1-amine hydrochloride

  • Cas Number : 1391443-56-3
  • Molecular Weight : 206.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-1-4-chloro-2-methylphenyl-ethan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87600
Availability details for
Quantity
  • Availability
  • Add to Cart