ADVH49B8327C AChemBlock

(R)-2-amino-2-(3-fluorophenyl)ethan-1-ol hydrochloride

  • Cas Number : 1391447-14-5
  • Molecular Weight : 191.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-2-amino-2-3-fluorophenyl-ethan-1-ol-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87601
Availability details for
Quantity
  • Availability
  • Add to Cart