ADVH49B83280 AChemBlock

(S)-2-(3-bromophenyl)pyrrolidine hydrochloride

  • Cas Number : 1391452-66-6
  • Molecular Weight : 262.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-2-3-bromophenyl-pyrrolidine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87605
Availability details for
Quantity
  • Availability
  • Add to Cart