ADVH49B83284 AChemBlock

(S)-1-(4-(trifluoromethoxy)phenyl)propan-1-amine hydrochloride

  • Cas Number : 1391484-89-1
  • Molecular Weight : 255.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-1-4-trifluoromethoxy-phenyl-propan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87609
Availability details for
Quantity
  • Availability
  • Add to Cart