ADVH49B83288 AChemBlock

(R)-1-(2,6-dichlorophenyl)propan-1-amine hydrochloride

  • Cas Number : 1391503-15-3
  • Molecular Weight : 240.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-1-2-6-dichlorophenyl-propan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87613
Availability details for
Quantity
  • Availability
  • Add to Cart