ADVH49B8328A AChemBlock

methyl (R)-3-(1-amino-2-hydroxyethyl)benzoate hydrochloride

  • Cas Number : 1391515-70-0
  • Molecular Weight : 231.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-r-3-1-amino-2-hydroxyethyl-benzoate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87615
Availability details for
Quantity
  • Availability
  • Add to Cart