ADVH49B83295 AChemBlock

2-bromo-4-chlorothiazole-5-carboxylic acid

  • Cas Number : 139670-04-5
  • Molecular Weight : 242.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-bromo-4-chlorothiazole-5-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87626
Availability details for
Quantity
  • Availability
  • Add to Cart