ADVH49B8329B AChemBlock

(S)-1-(2,3-difluorophenyl)ethan-1-amine hydrochloride

  • Cas Number : 1415303-43-3
  • Molecular Weight : 193.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-1-2-3-difluorophenyl-ethan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87632
Availability details for
Quantity
  • Availability
  • Add to Cart