ADVH49B832A5 AChemBlock

(R)-6-bromo-2,3-dihydro-1H-inden-1-amine hydrochloride

  • Cas Number : 1466429-22-0
  • Molecular Weight : 248.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-6-bromo-2-3-dihydro-1h-inden-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87642
Availability details for
Quantity
  • Availability
  • Add to Cart