ADVH49B832B0 AChemBlock

1-(6-chloropyridin-2-yl)ethan-1-one

  • Cas Number : 152356-57-5
  • Molecular Weight : 155.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-6-chloropyridin-2-yl-ethan-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87653
Availability details for
Quantity
  • Availability
  • Add to Cart