ADVH49B832B3 AChemBlock

cyclopropyl(pyridin-3-yl)methanol

  • Cas Number : 155047-86-2
  • Molecular Weight : 149.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/cyclopropyl-pyridin-3-yl-methanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87656
Availability details for
Quantity
  • Availability
  • Add to Cart