ADVH49B832B4 AChemBlock

(R)-1-(4-chlorophenyl)-N-methylethan-1-amine

  • Cas Number : 159053-43-7
  • Molecular Weight : 169.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-1-4-chlorophenyl-n-methylethan-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87657
Availability details for
Quantity
  • Availability
  • Add to Cart