ADVH49B832BD AChemBlock

6,7-dimethyl-2,3-dihydro-1H-inden-1-one

  • Cas Number : 16440-98-5
  • Molecular Weight : 160.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-7-dimethyl-2-3-dihydro-1h-inden-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87666
Availability details for
Quantity
  • Availability
  • Add to Cart