ADVH49B832BE AChemBlock

(S)-2-amino-2-(3-bromo-5-fluorophenyl)ethan-1-ol hydrochloride

  • Cas Number : 1646566-74-6
  • Molecular Weight : 270.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-2-amino-2-3-bromo-5-fluorophenyl-ethan-1-ol-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87667
Availability details for
Quantity
  • Availability
  • Add to Cart