ADVH49B832C3 AChemBlock

1-(pyridin-4-yl)butan-1-one

  • Cas Number : 1701-71-9
  • Molecular Weight : 149.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-pyridin-4-yl-butan-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87672
Availability details for
Quantity
  • Availability
  • Add to Cart