ADVH49B832C5 AChemBlock

1-(2-methoxyphenyl)ethan-1-amine hydrochloride

  • Cas Number : 17087-50-2
  • Molecular Weight : 187.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-2-methoxyphenyl-ethan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87674
Availability details for
Quantity
  • Availability
  • Add to Cart